missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dishevelled-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £360.00
Specifications
| Antigen | Dishevelled-1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 5-7 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18177641
|
Novus Biologicals
NBP2-24648SS |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18705313
|
Novus Biologicals
NBP2-24648 |
0.1 mg |
£360.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dishevelled-1 Polyclonal specifically detects Dishevelled-1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot.Specifications
| Dishevelled-1 | |
| Western Blot 5-7 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| O14640 | |
| 1855 | |
| A portion of amino acids 70-120 of human DVL-1 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction, Wnt Signaling Pathway | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| dishevelled 1 (homologous to Drosophila dsh), dishevelled, dsh homolog 1 (Drosophila), Dishevelled-1, DSH homolog 1, DVL, DVL1L1, MGC54245, segment polarity protein dishevelled homolog DVL-1 | |
| DVL1 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, pig, opossum, chimpanzee, dog and chicken, and 94% homologous in rat and mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title