missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Discoidin Domain Receptor Tyrosine Kinase 1 Rabbit Anti-Human, Polyclonal, Abnova&Trade;
Rabbit Polyclonal Antibody
Brand: Abnova PAB3365.400uL
This item is not returnable.
View return policy
Description
discoidin domain receptor tyrosine kinase 1 Polyclonal antibody specifically detects discoidin domain receptor tyrosine kinase 1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Flow CytometrySpecifications
| discoidin domain receptor tyrosine kinase 1 | |
| Polyclonal | |
| Flow Cytometry (1:10-50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| CAK/CD167/DDR/EDDR1/MCK10/NEP/NTRK4/PTK3/PTK3A/RTK6/TRKE | |
| A synthetic peptide (conjugated with KLH) corresponding to 24-54 amino acids at N-terminus of human DDR1. | |
| RUO | |
| 780 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| In PBS (0.09% sodium azide) | |
| Rabbit | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction