missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Discofix™-2 Two-Way Stopcock

Product Code. 11681379
Click to view available options
For Use With (Equipment):
Pressure Infusion Systems
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11681379

Brand: B Braun™ 4099010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A stopcock system for infusion therapy and monitoring.

  • Suitable for all pressure infusion systems used in infusion techniques, under 4 bar
  • Circulation holes for undistorted pressure valves
  • Axial and radial displacement coupling nut for safe and fast connection

Specifications

Color Blue
For Use With (Application) Infusion therapy and monitoring
Product Type Stopcock
Max. Pressure 4 bar
Quantity 50/Pk.
For Use With (Equipment) Pressure Infusion Systems
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.