Learn More
Hirschmann™ Discharge valve
Product Code. 10768291
Product Code. 10768291
Brand: Hirschmann™ 9316028
Only null left
Add to basket
Add to basket
Discharge valve are made of high purity alumina ceramics (Al2O3 99,7%). The spring is made of platin-iridium (HF).
  • For opus™, ceramus™, solarus™ & ceramus™ HF
Specifications
For Use With for opus™, ceramus™, solarus™, & ceramus™, HF