missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DISC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-76538-25ul
This item is not returnable.
View return policy
Description
DISC1 Polyclonal specifically detects DISC1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| DISC1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| C1orf136, disrupted in schizophrenia 1, disrupted in schizophrenia 1 protein, FLJ13381, FLJ21640, FLJ25311, FLJ41105, KIAA0457, SCZD9 | |
| Rabbit | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| DISC1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LQARMFVLEAKDQQLRREIEEQEQQLQWQGCDLTPLVGQLSLGQLQEVSKALQDTLASAGQIPFHAEPPETIRSLQERIKSLNLSLKEITTKVCMSEKFCSTLRKKVNDIET | |
| RUO | |
| 27185 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction