missing translation for 'onlineSavingsMsg'
Learn More

DISC1 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30734506 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734506 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734506 Supplier Novus Biologicals Supplier No. AF6699Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

DISC1 Polyclonal antibody specifically detects DISC1 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DISC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias C1orf136, disrupted in schizophrenia 1, disrupted in schizophrenia 1 protein, FLJ13381, FLJ21640, FLJ25311, FLJ41105, KIAA0457, SCZD9
Host Species Sheep
Immunogen E. coli-derived recombinant human DISC1, Lys101-Arg260, Accession # Q9NRI5
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cognition and Behavior, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 27185
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.