missing translation for 'onlineSavingsMsg'
Learn More

Disalicylimide, TRC

Product Code. 30388037
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388037 1 g -
30370628 100 mg 100mg
30371413 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388037 Supplier Toronto Research Chemicals Supplier No. TRCD4933051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1972-71-0
Molecular Formula C14 H11 N O4
Molecular Weight (g/mol) 257.2414
Synonym Benzamide, 2-hydroxy-N-(2-hydroxybenzoyl)-, Dibenzamide, 2,2'-dihydroxy- (6CI), Disalicylamide (7CI,8CI), 2-Hydroxy-N-(2-hydroxybenzoyl)benzamide, 2,2'-Dihydroxydibenzamide, 2-Hydroxy-N-[(2-hydroxyphenyl)carbonyl]benzamide, Disalicylimide, N-Salicyloylsalicylamide, Salicylimide, Deferasirox Imp. A (EP)
IUPAC Name 2-hydroxy-N-(2-hydroxybenzoyl)benzamide
SMILES Oc1ccccc1C(=O)NC(=O)c2ccccc2O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H11NO4/c16-11-7-3-1-5-9(11)13(18)15-14(19)10-6-2-4-8-12(10)17/h1-8,16-17H,(H,15,18,19)
Formula Weight 257.0688
Percent Purity >95%
Chemical Name or Material Disalicylamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.