missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIS3L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£222.00 - £451.00
Specifications
| Antigen | DIS3L2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18688205
|
Novus Biologicals
NBP2-38264-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18139628
|
Novus Biologicals
NBP2-38264 |
0.1 mL |
£451.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DIS3L2 Polyclonal specifically detects DIS3L2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DIS3L2 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| Q8IYB7 | |
| 129563 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RNRALNGDLVVVKLLPEEHWKVVKPESNDKETEAAYESDIPEELCGHHLPQQSLKSYNDSPDVIVEAQFDGSDSEDGHGITQNVLVDGVKKLS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DIS3 mitotic control homolog (S. cerevisiae)-like 2, DIS3-like exonuclease 2, FAM6A, member A | |
| DIS3L2 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title