missing translation for 'onlineSavingsMsg'
Learn More

DIS3L2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18407966 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407966 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407966 Supplier Novus Biologicals Supplier No. NBP281805AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DIS3L2 Polyclonal antibody specifically detects DIS3L2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIS3L2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias DIS3 mitotic control homolog (S. cerevisiae)-like 2, DIS3-like exonuclease 2, FAM6A, member A
Host Species Rabbit
Immunogen DIS3L2 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human DIS3L2. The immunogen is located within amino acids 720 - 770 of DIS3L2. Amino Acid Squence: DTLQKQADHCNDRRMAS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129563
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.