missing translation for 'onlineSavingsMsg'
Learn More

DIS3 Antibody, Novus Biologicals™

Product Code. 18368258 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368258 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368258 Supplier Novus Biologicals Supplier No. H00022894B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

DIS3 Polyclonal antibody specifically detects DIS3 in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIS3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10-1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH56143.1
Gene Alias 2810028N01Rik, DIS3 mitotic control homolog (S. cerevisiae), dis3p, DKFZp667L1817, EC 3.1.13, EC 3.1.13.-, EC 3.1.26.-, EXOSC11, exosome complex exonuclease RRP44, exosome component 11, FLJ10484, KIAA1008RP11-342J4.3, MGC33035, mitotic control protein dis3 homolog, Protein DIS3 homolog, Ribosomal RNA-processing protein 44, RRP44bA555G22.1
Host Species Mouse
Immunogen KIAA1008 (AAH56143.1, 1 a.a. - 958 a.a.) full-length human protein. MLKSKTFLKKTRAGGVMKIVREHYLRDDIGCGAPGCAACGGAHEGPALEPQPQDPASSVCPQPHYLLPDTNVLLHQIDVLEDPAIRNVIVLQTVLQEVRNRSAPVYKRIRDVTNNQEKHFYTFTNEHHRETYVEQEQGENANDRNDRAIRVAAKWYNEHLKKMSADNQLQVIFITNDRRNKEKAIEEGIPAFTCEEYVKSLTANPELIDRLACLSEEGNEIESGKIIFSEHLPLSKLQQGIKSGTYLQGTFRASRENYLEATVWIHGDNEENKEIILQGLKHLNRAVHEDIVAVELLPKSQWVAPSSVVLHDEGQNEEDVEKEEERERMLKTAVSEKMLKPTGRVVGIIKRNWRPYCGMLSKSDIKESRRHLFTPADKRIPRIRIETRQASTLEGRRIIVAIDGWPRNSRYPNGHFVRNLGDVGEKETETEVLLLEHDVPHQPFSQAVLSFLPKMPWSITEKDMKNREDLRHLCICSVDPPGCTDIDDALHCRELENGNLEVGVHIADVSHFIRPGNALDQESARRGTTVYLCEKRIDMVPELLSSNLCSLKCDVDRLAFSCIWEMNHNAEILKTKFTKSVINSKASLTYAEAQLRIDSANMNDDITTSLRGLNKLAKILKKRRIEKGALTLSSPEVRFHMDSETHDPIDLQTKELRETNSMVEEFMLLANISVAKKIHEEFSEHALLRKHPAPPPSNYEILVKAARSRNLEIKTDTAKSLAESLDQAESPTFPYLNTLLRILATRCMMQAVYFCSGMDNDFHHYGLASPIYTHFTSPIRRYADVIVHRLLAVAIGADCTYPELTDKHKLADICKNLNFRHKMAQYAQRASVAFHTQLFFKSKGIVSEEAYILFVRKNAIVVLIPKYGLEGTVFFEEKDKPNPQLIYDDEIPSLKIEDTVFHVFDKVKVKIMLDSSNLQHQKIRMSLVEPQIPGISIPTDTSNMDLNGPKKKKMKLGK
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 22894
Target Species Human, Mouse, Chicken
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.