missing translation for 'onlineSavingsMsg'
Learn More

DIS3 Antibody, Novus Biologicals™

Product Code. 18427791 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427791 25ul 25µL
18208427 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18427791 Supplier Novus Biologicals Supplier No. NBP18520925ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

DIS3 Polyclonal specifically detects DIS3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIS3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias 2810028N01Rik, DIS3 mitotic control homolog (S. cerevisiae), dis3p, DKFZp667L1817, EC 3.1.13, EC 3.1.13.-, EC 3.1.26.-, EXOSC11, exosome complex exonuclease RRP44, exosome component 11, FLJ10484, KIAA1008RP11-342J4.3, MGC33035, mitotic control protein dis3 homolog, Protein DIS3 homolog, Ribosomal RNA-processing protein 44, RRP44bA555G22.1
Gene Symbols DIS3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:EAYILFVRKNAIVVLIPKYGLEGTVFFEEKDKPNPQLIYDDEIPSLKIEDTVFHVFDKVKVKIMLDSSNLQHQKIRMSLVEPQIPGISIPTDTSN
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22894
Test Specificity Specificity of human DIS3 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.