missing translation for 'onlineSavingsMsg'
Learn More

DIS3 Antibody - BSA Free, Novus Biologicals™

Product Code. 18384101 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384101 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384101 Supplier Novus Biologicals Supplier No. NBP281804

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DIS3 Polyclonal specifically detects DIS3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIS3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 2810028N01Rik, DIS3 mitotic control homolog (S. cerevisiae), dis3p, DKFZp667L1817, EC 3.1.13, EC 3.1.13.-, EC 3.1.26.-, EXOSC11, exosome complex exonuclease RRP44, exosome component 11, FLJ10484, KIAA1008RP11-342J4.3, MGC33035, mitotic control protein dis3 homolog, Protein DIS3 homolog, Ribosomal RNA-processing protein 44, RRP44bA555G22.1
Gene Symbols DIS3
Host Species Rabbit
Immunogen DIS3 antibody was raised against an 18 amino acid peptide near the amino terminus of human DIS3. The immunogen is located within amino acids 90 - 140 of DIS3. Amino Acid Squence: YKRIRDVTNNQEKHFYTF
Molecular Weight of Antigen 103 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22894
Test Specificity Multiple isoforms of DIS3 are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.