missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diroximel fumarate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Diroximel fumarate (ALKS 8700) is an orally-active and well-tolerated monomethyl fumarate (MMF) prodrug in a controlled-release formulation. Diroximel fumarate is considered as active equivalent to its active metabolite dimethyl fumarate (DMF). Diroximel fumarate has a favorable safety and efficacy profile, has the potential for the study of multiple sclerosis (MS).
Specifications
Specifications
| Chemical Name or Material | Diroximel fumarate |
| CAS | 1577222-14-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C11H13NO6 |
| Quantity | 5 mg |
| Synonym | ALKS 8700 BIIB098 |
| Solubility Information | DMSO : 125 mg/mL (489.77 mM; Need ultrasonic) |
| SMILES | O=C(OCCN1C(CCC1=O)=O)/C=C/C(OC)=O |
| Molecular Weight (g/mol) | 255.22 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction