missing translation for 'onlineSavingsMsg'
Learn More

Dirlotapide, MedChemExpress

Product Code. 16423129
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423129 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16423129 Supplier Medchem Express Supplier No. HYU00070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dirlotapide (CP742033) is a gut-selective inhibitor of microsomal triglyceride transfer protein (MTP) that reliably produces weight loss in obese dogs.

Dirlotapide (CP742033) is a gut-selective inhibitor of microsomal triglyceride transfer protein (MTP) that reliably produces weight loss in obese dogs.

Specifications

Chemical Name or Material Dirlotapide
CAS 481658-94-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C40H33F3N4O3
Quantity 1 mg
Synonym CP742033 Slentrol
SMILES O=C(C(N1C)=CC2=C1C=CC(NC(C3=CC=CC=C3C4=CC=C(C(F)(F)F)C=C4)=O)=C2)N[C@@H](C5=CC=CC=C5)C(N(C)CC6=CC=CC=C6)=O
Molecular Weight (g/mol) 674.71
Formula Weight 674.71
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.