missing translation for 'onlineSavingsMsg'
Learn More

Dirithromycin, TRC

Product Code. 30136923
Change view
Click to view available options
Quantity:
5 mg
100 mg
10 g
Unit Size:
100mg
10g
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136923 10 g 10g
30136809 100 mg 100mg
17930775 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136923 Supplier Toronto Research Chemicals Supplier No. TRCD49300010G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62013-04-1
Molecular Formula C42 H78 N2 O14
Molecular Weight (g/mol) 835.07
Synonym Dirithromycin, 1R,2S,3R,6R,7S,8S,9R,10R,12R,13S,15R,17S)-9-[[3-(Dimethylamino)-3,4,6-trideoxy-β-d-xylo-hexopyranosyl]oxy]-3-ethyl-2,10-dihydroxy-15-[(2-methoxyethoxy)methyl]-2,6,8,10,12,17-hexamethyl-7-[(3-C-methyl-3-O-methyl-2,6dideoxy-α-l-ribo-hexopyranosyl)oxy]-4,16-dioxa-14azabicyclo[11.3.1]heptadecan-5-one, (9S)-9,11-[imino[(1R)-2-(2-methoxyethoxy)ethylidene]oxy]-9-deoxo-11-deoxyerythromycin
IUPAC Name (1S,2R,4R,5R,6S,7S,8R,11R,12R,13R,15R)-5-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-11-ethyl-4,12-dihydroxy-7-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-15-(2-methoxyethoxymethyl)-2,4,6,8,12,17-hexamethyl-10,14-dioxa-16-azabicyclo[11.3.1]heptadecan-9-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)[C@@H]4N[C@@H](COCCOC)O[C@H](C4C)[C@]1(C)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C42H78N2O14/c1-15-29-42(10,49)37-24(4)32(43-30(56-37)21-52-17-16-50-13)22(2)19-40(8,48)36(58-39-33(45)28(44(11)12)18-23(3)53-39)25(5)34(26(6)38(47)55-29)57-31-20-41(9,51-14)35(46)27(7)54-31/h22-37,39,43,45-46,48-49H,15-21H2,1-14H3/t22-,23-,24?,25+,26-,27+,28+,29-,30-,31+,32+,33-,34+,35+,36-,37-,39+,40-,41-,42-/m1/s1
Formula Weight 834.5453
Percent Purity >95%
Chemical Name or Material Dirithromycin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.