missing translation for 'onlineSavingsMsg'
Learn More

Dirithromycin, MedChemExpress

Product Code. 16402259
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402259 1 g 1g
16401038 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16402259 Supplier Medchem Express Supplier No. HYB0643
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dirithromycin (LY237216), a derivative of Erythromycin, is a potent and orally active semi-synthetic macrolide antibiotic. Dirithromycin is active against gram-positive bacteria, Legionella spp., Helicobacter pylori, and Chlamydia trachomatis.

Dirithromycin (LY237216), a derivative of Erythromycin, is a potent and orally active semi-synthetic macrolide antibiotic. Dirithromycin is active against gram-positive bacteria, Legionella spp., Helicobacter pylori, and Chlamydia trachomatis.

Spécification

Chemical Name or Material Dirithromycin
Physical Form Solid
CAS 62013-04-1
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C42H78N2O14
For Use With (Application) COVID-19-immunoregulation
Synonym LY237216
Solubility Information Ethanol : ≥ 50 mg/mL (59.88 mM) ∣DMSO : 33.33 mg/mL (39.91 mM; Need ultrasonic)
SMILES CC1[C@@]([C@](O)([C@H](OC([C@@H]2C)=O)CC)C)([H])O[C@H](COCCOC)N[C@@]1([H])[C@](C[C@@](O)([C@@H]([C@@H](C)[C@]2([H])O[C@@](O[C@@H](C)[C@@H]3O)([H])C[C@@]3(C)OC)O[C@@](O[C@H](C)C[C@@H]4N(C)C)([H])[C@@H]4O)C)([H])C
Molecular Weight (g/mol) 835.07
Formula Weight 835.07
Percent Purity 98.0%
Grade Research
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.