missing translation for 'onlineSavingsMsg'
Learn More

DiversifiedBiotech Direct Thermal CRYO-TAGS™ Roll Labels
GREENER_CHOICE

Product Code. 30282341
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1000/Pk.
750/Pk.
Width (English):
0.50 in.
0.75 in.
missing translation for 'unitSize'
Pack of 1000
Pack of 750
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Width (English) unitSize
30282341 1000/Pk. 0.50 in. Pack of 1000
30282307 750/Pk. 0.50 in. Pack of 750
30282275 750/Pk. 0.75 in. Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Product Code. 30282341 missing translation for 'mfr' DiversifiedBiotech missing translation for 'supplierNo' DTCR1000
Only null left
Add to basket
Add to basket

For quick label samples with sharp text, clear barcodes

  • Direct Thermal Cryo-Tags™ formatted to work with most direct thermal printers
  • Quickly label your samples with sharp text, clear barcodes or a combination of both
  • Cryogenic labels are suitable for a variety of applications
  • Diversified Biotech recommends the Label LIVE app for printing in conjunction with the mydpi Label Printer

Specifications

Adhesive Type Peel and Stick, Acrylic
Color White
Finish Semi-gloss
Format Roll
Material Polyolefin
Print Technology Direct Thermal
Product Type Ultra Reflective Letter Label
Sterility Non-sterile
Quantity 1000/Pk.
Dimensions (L x W) 1.05 x 0.50 in.
Length (English) 1.05 in.
Width (English) 0.50 in.
Width (Metric) 1.27 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.