missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Direct Red 80

Product Code. 11309739
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11309739

Brand: Thermo Scientific Chemicals B21693.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 2610-10-8 | C45H26N10Na6O21S6 | 1373.05 g/mol

Direct red 80 is a dye useful as a cardiac muscle stain and as a model compound in adsorption studies. It has also been utilized to compare efficacy of textile dye removal in aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Direct red 80 is a dye useful as a cardiac muscle stain and as a model compound in adsorption studies. It has also been utilized to compare efficacy of textile dye removal in aqueous solutions.

Solubility
Partly soluble in water, very slightly soluble in ethanol, and cellosolve. Insoluble in many organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2610-10-8
MDL Number MFCD00054389
Synonym C.I. 35780; Sirius Red

Specifications

Molecular Formula C8H8O4
Solubility Information Partly soluble in water,very slightly soluble in ethanol,and cellosolve. Insoluble in many organic solvents.
InChI Key IZZIWIAOVZOBLF-UHFFFAOYSA-N
SMILES COC1=CC(=C(C=C1)O)C(=O)O
IUPAC Name 2-hydroxy-5-methoxybenzoic acid
Molecular Weight (g/mol) 168.15
PubChem CID 75787
Formula Weight 1373.09
Quantity 25 g
Chemical Name or Material Direct Red 80

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.