missing translation for 'onlineSavingsMsg'
Learn More

Direct Brown 95 (Technical Grade), TRC

Product Code. 30007029
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007029 1 g 1g
30011944 500 mg 500mg
30012588 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007029 Supplier Toronto Research Chemicals Supplier No. TRCD4930201G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16071-86-6
Molecular Formula C31 H18 Cu N6 O9 S . 2 Na
Molecular Weight (g/mol) 760.1
Synonym Cuprate(2-), [2-hydroxy-5-[2-[4'-[2-[2-(hydroxy-κO)-6-hydroxy-3-[2-[2-(hydroxy-κO)-5-sulfophenyl]diazenyl-κN1]phenyl]diazenyl][1,1'-biphenyl]-4-yl]diazenyl]benzoato(4-)]-, sodium (1:2), C.I. Direct Brown 95 (7CI), Copper, [dihydrogen 5-[[4'-[[2,6-dihydroxy-3-[(2-hydroxy-5-sulfophenyl)azo]phenyl]azo]-4-biphenylyl]azo]salicylato(2-)]-, disodium salt (8CI), Cuprate(2-), [5-[[4'-[[2,6-dihydroxy-3-[(2-hydroxy-5-sulfophenyl)azo]phenyl]azo][1,1'-biphenyl]-4-yl]azo]-2-hydroxybenzoato(4-)]-, disodium, Cuprate(2-), [5-[[4'-[[2-(hydroxy-κO)-6-hydroxy-3-[[2-(hydroxy-κO)-5-sulfophenyl]azo-κN1]phenyl]azo][1,1'-biphenyl]-4-yl]azo]-2-hydroxybenzoato(4-)]-, disodium (9CI), Benzoic acid, 5-[[4'-[[2,6-dihydroxy-3-[(2-hydroxy-5-sulfophenyl)azo]phenyl]azo][1,1'-biphenyl]-4-yl]azo]-2-hydroxy-, copper complex, Aizen Primula Brown BRLH, Aizen Primula Brown PLH, Amanil Fast Brown BRL, Amanil Supra Brown LBL, Atlantic Fast Brown BRL, Atlantic Resin Fast Brown BRL, Belamine Fast Brown BRLL, Benzanil Supra Brown BRLL, Benzanil Supra Brown BRLN, Brown 4EMBL, C.I. 30145, Calcodur Brown BRL, Chloramine Fast Brown BRL, Chloramine Fast Cutch Brown PL, Chlorantine Fast Brown BRLL, Chrome Leather Brown BRLL, Chrome Leather Brown BRSL, Cuprofix Brown GL, Derma Fast Brown W-GL, Dermafix Brown PL, Dialuminous Brown BRS, Diaphtamine Light Brown BRLL, Diazine Fast Brown RSL, Diazol Light Brown BRN, Dicorel Brown LMR, Diphenyl Fast Brown BRL, Direct Brown 95, Direct Brown BRL, Direct Fast Brown BRL, Direct Fast Brown LMR, Direct Light Brown BRS, Direct Lightfast Brown M, Direct Supra Light Brown ML, Durazol Brown BR, Durofast Brown BRL, Eliamina Light Brown BRL, Enianil Light Brown BRL, Fastolite Brown BRL, Fastusol Brown LBRSA, Fastusol Brown LBRSN, Fenaluz Brown BRL, Helion Brown BRSL, Hispaluz Brown BRL, KCA Light Fast Brown BR, Kayarus Supra Brown BRS, Paranol Fast Brown BRL, Peeramine Fast Brown BRL, Pontamine Fast Brown BRL, Pontamine Fast Brown NP, Pyrazol Fast Brown BRL, Pyrazoline Brown BRL, Saturn Brown LBR, Sirius Supra Brown BR, Sirius Supra Brown BRL, Sirius Supra Brown BRS, Solantine Brown BRL, Solar Brown PL, Solex Brown R, Solius Light Brown BRLL, Solius Light Brown BRS, Sumilight Supra Brown BRS, Suprazo Brown BRL, Suprexcel Brown BRL, Tertrodirect Fast Brown BR, Tetramine Fast Brown BRDN Extra, Tetramine Fast Brown BRP, Tetramine Fast Brown BRS, Triantine Brown BRS, Triantine Fast Brown OG, Triantine Fast Brown OR, Triantine Light Brown BRS, Triantine Light Brown OG, Vopsider Brown DLMR
IUPAC Name Direct Brown 95
SMILES [Na+].[Na+].Oc1ccc(cc1C(=O)[O-])N=Nc2ccc(cc2)c3ccc(cc3)N=Nc4c(O)ccc5c4[O-][Cu+2]6N5=Nc7cc(ccc7[O-]6)S(=O)(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C31H22N6O9S.Cu.2Na/c38-26-12-9-21(15-23(26)31(42)43)34-32-19-5-1-17(2-6-19)18-3-7-20(8-4-18)33-37-29-28(40)14-11-24(30(29)41)35-36-25-16-22(47(44,45)46)10-13-27(25)39;;;/h1-16,38-41H,(H,42,43)(H,44,45,46);;;/q;+2;2*+1/p-4/b34-32+,36-35+,37-33+;;;
Formula Weight 758.9947
Chemical Name or Material Direct Brown 95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.