missing translation for 'onlineSavingsMsg'
Learn More

Direct Blue 76 (Technical Grade), TRC

Product Code. 30010510
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010510 100 mg 100mg
30012740 250 mg 250mg
30010037 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010510 Supplier Toronto Research Chemicals Supplier No. TRCD492955100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16143-79-6
Molecular Formula C32H16Cu2N6O16S4.4Na
Molecular Weight (g/mol) 1087.82
Synonym 1,3-Naphthalenedisulfonic acid, 6,6'-[(3,3'-dihydroxy[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[4-amino-5-hydroxy-, Copper Complex, Benzanil Supra Blue 2GN, C.I. 24411, C.I. Direct Blue 76, Chlorantine Fast, Blue 7GL, Chlorantine Fast Blue 7GLL, Direct Blue 76, Durazol Blue 2GN, Durazol Blue 2GNP, Lumicrease Blue 4GL, Lumicrease Sky Blue 4GL, Lumicrease Sky Blue, 6GUL, Pontamine Fast Blue 6GL, Pyrazol Fast Blue 4GUL, Sirius Blue F4GA;
IUPAC Name 1,3-Naphthalenedisulfonic acid, 6,6'-[(3,3'-dihydroxy[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[4-amino-5-hydroxy-, Copper Complex; Benzanil Supra Blue 2GN; C.I. 24411; C.I. Direct Blue 76; Chlorantine Fast; Blue 7GL; Chlorantine Fast Blue 7GLL; Direct Blue 76; Durazol Blue 2GN; Durazol Blue 2GNP; Lumicrease Blue 4GL; Lumicrease Sky Blue 4GL; Lumicrease Sky Blue; 6GUL; Pontamine Fast Blue 6GL; Pyrazol Fast Blue 4GUL; Sirius Blue F4GA;
SMILES [Na+].O=S(=O)([O-])C=1C=C(C=2C=CC3=C([O-][Cu+2]4[O-]C=5C=C(C=CC5N=[N]34)C6=CC=C7N=[N]8C=9C=CC%10=C(C9[O-][Cu+2]8[O-]C7=C6)C(N)=C(C=C%10S(=O)(=O)[O-])S(=O)(=O)[O-])C2C1N)S(=O)(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChi=1S/C32H24N6O16S4.2Cu.4Na/c33-29-25(57(49,50)51)11-23(55(43,44)45)15-3-7-19(31(41)27(15)29)37-35-17-5-1-13(9-21(17)39)14-2-6-18(22(40)10-14)36-38-20-8-4-16-24(56(46,47)48)12-26(58(52,53)54)30(34)28(16)32(20)42;;;;;;/h1-12,39-42H,33-34H2,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;;;/q;2*+2;4*+1/p-8
Formula Weight 1085.77
Chemical Name or Material Direct Blue 76
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.