missing translation for 'onlineSavingsMsg'
Learn More

Direct Blue 6 (Technical Grade), TRC

Product Code. 30040572
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040572 250 mg 250mg
30040857 1 g 1g
18063820 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040572 Supplier Toronto Research Chemicals Supplier No. TRCD492950250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2602-46-2
Molecular Formula C32 H20 N6 O14 S4 . 4 Na
Molecular Weight (g/mol) 932.75
Synonym 1-Naphthol-3,6-disulfonic acid, 2,2'-(4,4'-biphenylylenebisazo)bis[8-amino-, tetrasodium salt (6CI), 2,7-Naphthalenedisulfonic acid, 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis[5-amino-4-hydroxy-, tetrasodium salt (9CI), C.I. Direct Blue 6 (7CI), C.I. Direct Blue 6, tetrasodium salt (8CI), Airedale Blue 2BD, Aizen Direct Blue 2BH, Amanil Blue 2BX, Atlantic Blue 2B, Atul Direct Blue 2B, Azocard Blue 2B, Azomine Blue 2B, Belamine Blue 2B, Bencidal Blue 2B, Benzanil Blue 2B, Benzo Blue 2B, Benzo Blue BBA-CF, Benzo Blue BBN-CF, Benzo Blue GS, Blue 2B, Blue 2B salt, Brasilamina Blue 2B, C.I. 22610, Calcomine Blue 2B, Chloramine Blue 2B, Chlorazol Blue B, Chlorazol Blue BP, Chrome Leather Blue 2B, Cresotine Blue 2B, Diacotton Blue BB, Diamine Blue, Diamine Blue 2B, Diamine Blue BB, Diaphtamine Blue BB, Diazine Blue 2B, Diazol Blue 2B, Diphenyl Blue 2B, Diphenyl Blue KF, Diphenyl Blue M 2B, Direct Blue 2B, Direct Blue 2BA, Direct Blue 6, Direct Blue A, Direct Blue BB, Direct Blue GS, Direct Blue K, Direct Blue M 2B, Direct Sky Blue K, Enianil Blue 2BN, Fenamin Blue 2B, Fixanol Blue 2B, Hispamin Blue 2B, Indigo Blue 2B, Kayaku Direct Blue BB, Mitsui Direct Blue 2BN, Naphtamine Blue 2B, Niagara Blue 2B, Nippon Blue BB, Paramine Blue 2B, Phenamine Blue BB, Pheno Blue 2B, Pontamine Blue BB, Tertrodirect Blue 2B, Vondacel Blue 2B, 2,7-Naphthalenedisulfonic acid, 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(2,1-diazenediyl)]bis[5-amino-4-hydroxy-, sodium salt (1:4) (ACI), Direct Blue 2b
IUPAC Name tetrasodium;5-amino-3-[(E)-[4-[4-[(E)-(8-amino-1-hydroxy-3,6-disulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]-4-hydroxynaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].Nc1cc(cc2cc(c(N=Nc3ccc(cc3)c4ccc(cc4)N=Nc5c(O)c6c(N)cc(cc6cc5S(=O)(=O)[O-])S(=O)(=O)[O-])c(O)c12)S(=O)(=O)[O-])S(=O)(=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C32H24N6O14S4.4Na/c33-23-13-21(53(41,42)43)9-17-11-25(55(47,48)49)29(31(39)27(17)23)37-35-19-5-1-15(2-6-19)16-3-7-20(8-4-16)36-38-30-26(56(50,51)52)12-18-10-22(54(44,45)46)14-24(34)28(18)32(30)40;;;;/h1-14,39-40H,33-34H2,(H,41,42,43)(H,44,45,46)(H,47,48,49)(H,50,51,52);;;;/q;4*+1/p-4/b37-35+,38-36+;;;;
Formula Weight 931.9511
Chemical Name or Material Direct Blue 2b
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.