missing translation for 'onlineSavingsMsg'
Learn More

Direct Black 168 (tech grade), TRC

Product Code. 30387038
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387038 50 mg
30378150 25 mg
30371966 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387038 Supplier Toronto Research Chemicals Supplier No. TRCD49482050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85631-88-5
Molecular Formula C34 H24 N9 Na3 O11 S3
Molecular Weight (g/mol) 899.772
Synonym 4-amino-3-[2-[4-[[4-[2-(2-amino-4-hydroxyphenyl)diazenyl]phenyl]amino]-3-sulfophenyl]diazenyl]-5-hydroxy-6-(2-phenyldiazenyl)-2,7-Naphthalenedisulfonic acid sodium salt, 4-amino-3-[[4-[[4-[(2-amino-4-hydroxyphenyl)azo]phenyl]amino]-3-sulfophenyl]azo]-5-hydroxy-6-(phenylazo)-2,7-Naphthalenedisulfonic acid trisodium salt, C.I. 335475, C.I. Direct Black 168, Coriacide Black SB, Derma Carbon NBS, Direct Black H 8F-SF-VP132, Direct Black HEF Liq., Direct Black HEF-SF, Duasyn Black HEF-SF Liquid, Duasyn Direct Black HEF, Duasyn Direct Black HEF-SF
IUPAC Name [(3E)-5-amino-6-[(E)-[4-[4-[(2E)-2-(2-amino-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]anilino]-3-sodiooxysulfonylphenyl]diazenyl]-4-oxo-3-(phenylhydrazinylidene)-7-sodiooxysulfonylnaphthalen-2-yl]sulfonyloxysodium
SMILES NC1=CC(=O)C=C/C/1=N\Nc2ccc(Nc3ccc(cc3S(=O)(=O)O[Na])\N=N\c4c(N)c5C(=O)\C(=N/Nc6ccccc6)\C(=Cc5cc4S(=O)(=O)O[Na])S(=O)(=O)O[Na])cc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C34H27N9O11S3.3Na/c35-24-17-23(44)11-13-25(24)41-38-21-8-6-19(7-9-21)37-26-12-10-22(16-27(26)55(46,47)48)40-42-32-28(56(49,50)51)14-18-15-29(57(52,53)54)33(34(45)30(18)31(32)36)43-39-20-4-2-1-3-5-20;;;/h1-17,37-39H,35-36H2,(H,46,47,48)(H,49,50,51)(H,52,53,54);;;/q;3*+1/p-3/b41-25+,42-40+,43-33-;;;
Formula Weight 899.045
Chemical Name or Material Direct Black 168 (tech grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.