missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIRC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100780
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DIRC1.
This gene consists of two exons and encodes a 104aa protein. Disruption of this gene by translocation t(2;3)(q33;q21) is associated with familial clear cell renal cancer.
Specifications
| DIRC1 | |
| Polyclonal | |
| Unconjugated | |
| DIRC1 | |
| DIRC1; disrupted in renal cancer protein; disrupted in renal carcinoma 1; disrupted in renal carcinoma protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116093 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q969H9 | |
| DIRC1 | |
| A synthesized peptide derived from human DIRC1(Accession Q969H9), corresponding to amino acid residues N48-L98. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction