missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIRAS3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555898
This item is not returnable.
View return policy
Description
Immunogen sequence: MGNASFGSKE QKLLKRLRLL PALLILRAFK PHRKIRDYRV VVVGTAGVGK STLLHKWASG NFRHEYLP Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 29%.
DIRAS3 Polyclonal antibody specifically detects DIRAS3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications
| DIRAS3 | |
| Polyclonal | |
| Unconjugated | |
| DIRAS3 | |
| ARHI; DIRAS family GTP binding RAS like 3; DIRAS family GTPase 3; DIRAS family, GTP-binding RAS-like 3; DIRAS3; Distinct subgroup of the Ras family member 3; GTP-binding protein Di-Ras3; NOEY2; ras homolog gene family, member I; RGD1565168; RHOI; rho-related GTP-binding protein RhoI | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9077 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95661 | |
| DIRAS3 | |
| Recombinant protein corresponding to Human DIRAS3 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction