Learn More
Invitrogen™ DIRAS1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101620
Product Code. 16678094
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total DIRA1.
DIRAS1 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIRAS1
Polyclonal
Unconjugated
Diras1
DIRAS family GTP binding RAS like 1; DIRAS family GTPase 1; DIRAS family, GTP-binding RAS-like 1; DIRAS1; Di-Ras1; Distinct subgroup of the Ras family member 1; GBTS1; GTP-binding protein Di-Ras1; ras-related inhibitor of cell growth; RIG; small GTP-binding tumor suppressor 1
Rabbit
Affinity Chromatography
RUO
148252
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95057
Diras1
A synthesized peptide derived from human DIRAS1(Accession O95057), corresponding to amino acid residues F161-M198.