missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIRAS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104509
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DIRA1.
DIRAS1 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.
Specifications
| DIRAS1 | |
| Polyclonal | |
| Unconjugated | |
| Diras1 | |
| DIRAS family GTP binding RAS like 1; DIRAS family GTPase 1; DIRAS family, GTP-binding RAS-like 1; DIRAS1; Di-Ras1; Distinct subgroup of the Ras family member 1; GBTS1; GTP-binding protein Di-Ras1; ras-related inhibitor of cell growth; RIG; small GTP-binding tumor suppressor 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 148252, 208666 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95057, Q91Z61 | |
| Diras1 | |
| A synthesized peptide derived from human DIRAS1(Accession O95057), corresponding to amino acid residues F161-M198. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction