missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIRAS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58888
This item is not returnable.
View return policy
Description
DIRAS1 Polyclonal specifically detects DIRAS1 in Human samples. It is validated for Western Blot.
Specifications
| DIRAS1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DIRAS family, GTP-binding RAS-like 1, Di-Ras1, Distinct subgroup of the Ras family member 1, GBTS1Small GTP-binding tumor suppressor 1, GTP-binding protein Di-Ras1, Ras-related inhibitor of cell growth, Rig, RIGFLJ42681 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 92%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Yeast, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O95057 | |
| DIRAS1 | |
| Synthetic peptides corresponding to DIRAS1(DIRAS family, GTP-binding RAS-like 1) The peptide sequence was selected from the N terminal of DIRAS1. Peptide sequence PEQSNDYRVVVFGAGGVGKSSLVLRFVKGTFRDTYIPTIEDTYRQVISCD. | |
| 100 μL | |
| Signal Transduction | |
| 148252 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction