missing translation for 'onlineSavingsMsg'
Learn More

Dipyrocetyl, MedChemExpress

Product Code. 16498678
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498678 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498678 Supplier Medchem Express Supplier No. HYB1179
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dipyrocetyl is an anti-inflammatory and analgesic agent extracted from patent WO 2011132171 A1.

Dipyrocetyl is an anti-inflammatory and analgesic agent extracted from patent WO 2011132171 A1.

Specifications

Chemical Name or Material Dipyrocetyl
CAS 486-79-3
Color White
Purity Grade Notes Research
Molecular Formula C11H10O6
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (419.83 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC(OC(C)=O)=C1OC(C)=O
Molecular Weight (g/mol) 238.19
Formula Weight 238.19
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.