missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipyridamole EP Impurity C, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Dipyridamole EP Impurity C
Specifications
Specifications
| Chemical Name or Material | 2,2'-((6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl)nitrilo)diethanol |
| CAS | 54093-92-4 |
| Analyte or Component Names | 2,2'-((6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl)nitrilo)diethanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H30 Cl N7 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2,2'-[[6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]nitrilo]diethanol, Dipyridamole Imp. C (EP), Dipyridamole USP Related Compound C, Dipyridamole USP RC C, 2,2'-[[6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]nitrilo]diethanol, Dipyridamole Related Compound C, Dipyridamole Impurity C |
| InChI Formula | InChI=1S/C20H30ClN7O2/c21-19-22-15-16(17(24-19)26-7-3-1-4-8-26)23-20(28(11-13-29)12-14-30)25-18(15)27-9-5-2-6-10-27/h29-30H,1-14H2 |
| SMILES | OCCN(CCO)c1nc(N2CCCCC2)c3nc(Cl)nc(N4CCCCC4)c3n1 |
| IUPAC Name | 2-[[2-chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction