missing translation for 'onlineSavingsMsg'
Learn More

Melford DIPSO

Product Code. 30018243
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018243 25 g 25g
30018242 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018243 Supplier Melford Supplier No. D4408025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Substance Name: 3-(N,N-Bis(2-Hydroxyethyl)amino)-2-hydroxypropane sulfonic acid

CAS Number: 68399-80-4

Molecular Formula: C7H17NO6S

Molecular Weight: 243.3

Chemical Purity: >98%

Soluble in:Water

Store at: Room Temperature

Specifications

Chemical Name or Material DIPSO
CAS 68399-80-4
Quantity 25 g
Molecular Formula C7H17NO6S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.