missing translation for 'onlineSavingsMsg'
Learn More

Dipsacoside B, MedChemExpress

Product Code. 16431368
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431368 5 mg 5mg
16428928 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431368 Supplier Medchem Express Supplier No. HYN0266
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dipsacoside B is a major bioactive saponin, which can be used as a marker.

Dipsacoside B is a major bioactive saponin, which can be used as a marker.

Specifications

Chemical Name or Material Dipsacoside B
CAS 33289-85-9
Color White
Purity Grade Notes Research
Molecular Formula C53H86O22
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (93.00 mM; Need ultrasonic)
SMILES O=C([C@]1(CCC(C)(C)C2)[C@]2([H])C3=CC[C@@]4([H])[C@@](C)(CC[C@]5([H])[C@@]4(CC[C@H](O[C@@](OC[C@H](O)[C@@H]6O)([H])[C@@H]6O[C@@](O[C@@H](C)[C@H](O)[C@H]7O)([H])[C@@H]7O)[C@@]5(C)CO)C)[C@]3(C)CC1)O[C@@H]([C@@H]([C@@H](O)[C@@H]8O)O)O[C@@H]8CO[C@@H]([C@@H]([C@@H](O)[C@@H]9O)O)O[C@@H]9CO
Molecular Weight (g/mol) 1075.24
Formula Weight 1075.24
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.