missing translation for 'onlineSavingsMsg'
Learn More

Dipropylamine, 99%

Product Code. 10599420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10599420 1 L Glass bottle 1 L
10488830 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10599420 Supplier Thermo Scientific Acros Supplier No. 117490010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 142-84-7 | C6H15N | 101.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-84-7
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00009362
InChI Key WEHWNAOGRSTTBQ-UHFFFAOYSA-N
Synonym dipropylamine, di-n-propylamine, 1-propanamine, n-propyl, n-dipropylamine, n-propyl-1-propanamine, rcra waste number u110, dipropyl amine, rcra waste no. u110, unii-60p318iiry, ccris 4805
PubChem CID 8902
IUPAC Name N-propylpropan-1-amine
SMILES CCCNCCC

Specifications

Melting Point -63.0°C
Density 0.7400g/mL
Boiling Point 105.0°C to 110.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CH2CH2)2NH
Refractive Index 1.4030 to 1.4050
Quantity 1 L
Beilstein 04, 138
Merck Index 15, 3383
Specific Gravity 0.74
Solubility Information Solubility in water: 46g/L (20°C). Other solubilities: freely soluble in alcohol
Viscosity 0.5 mPa.s (20°C)
Formula Weight 101.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dipropylamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dipropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.