missing translation for 'onlineSavingsMsg'
Learn More

Dipraglurant, MedChemExpress

Product Code. 16481449
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481449 1 mg 1mg
16448807 5 mg 5mg
16412347 50 mg 50mg
16406218 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481449 Supplier Medchem Express Supplier No. HY14859
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dipraglurant (ADX48621) is a potent, selective, orally active and brain penetrant mGluR5 negative allosteric modulator (NAM), with an IC50 of 21 nM. Dipraglurant can reduce Levodopa-induced dyskinesia (LID) in vivo.

Dipraglurant (ADX48621) is a potent, selective, orally active and brain penetrant mGluR5 negative allosteric modulator (NAM), with an IC50 of 21 nM. Dipraglurant can reduce Levodopa-induced dyskinesia (LID) in vivo.

Specifications

Chemical Name or Material Dipraglurant
CAS 872363-17-2
Color White
Purity Grade Notes Research
Molecular Formula C16H12FN3
Quantity 1 mg
Synonym ADX48621
Solubility Information DMSO : ≥ 40 mg/mL (150.78 mM)
SMILES FC1=CN2C(C=C1)=NC(CCC#CC3=NC=CC=C3)=C2
Molecular Weight (g/mol) 265.29
Formula Weight 265.29
Percent Purity 95.95%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.