missing translation for 'onlineSavingsMsg'
Learn More

Dipotassium Glycyrrhizinate (~80%), TRC

Product Code. 30384852
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384852 10 mg
30373951 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384852 Supplier Toronto Research Chemicals Supplier No. TRCD49214510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68797-35-3
Molecular Formula C42H62K2O16
Molecular Weight (g/mol) 901.13
Synonym (3β,20β)-20-Carboxy-11-oxo-30-norolean-12-en-3-yl 2-O-β-D-glucopyranuronosyl-α-D-glucopyranosiduronic Acid Dipotassium Salt, 18β-Glycyrrhizic Acid Dipotassium Salt, Dipotassium Glycyrrhizate, Glycyrrhizic Acid Dipotassium Salt, Glycyrrhizin Dipotassium Salt, Glycyrrhizinic Acid Dipotassium Salt, Licozin G 1, OriStar DPG, (2S,3S,4S,5R,6R)-6-(((2S,3R,4S,5S,6S)-6-Carboxy-2-(((3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl)oxy)-4,5-dihydroxytetrahydro-2H-pyran-3-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic Acid Dipotassium Salt
IUPAC Name dipotassium;(2S,3S,4S,5R,6S)-6-[[(3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxylato-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-2,3,4a,5,6,7,8,9,10,12,12a,14a-dodecahydro-1H-picen-3-yl]oxy]-5-[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy-3,4-dihydroxyoxane-2-carboxylate
SMILES [H][C@@]12C[C@@](C)(C(O)=O)CC[C@]1(C)CC[C@]3(C)C2=CC([C@@]4([H])[C@@]3(C)CC[C@]5([H])[C@]4(C)CC[C@H](O[C@H]6O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]6O[C@@H]7[C@@H](O)[C@H](O)[C@@H](O)[C@H](C(O)=O)O7)C5(C)C)=O.[2 K]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C42H62O16.2K/c1-37(2)21-8-11-42(7)31(20(43)16-18-19-17-39(4,36(53)54)13-12-38(19,3)14-15-41(18,42)6)40(21,5)10-9-22(37)55-35-30(26(47)25(46)29(57-35)33(51)52)58-34-27(48)23(44)24(45)28(56-34)32(49)50;;/h16,19,21-31,34-35,44-48H,8-15,17H2,1-7H3,(H,49,50)(H,51,52)(H,53,54);;/q;2*+1/p-2/t19-,21-,22-,23-,24-,25-,26-,27+,28-,29-,30+,31+,34-,35-,38+,39-,40-,41+,42+;;/m0../s1
Formula Weight 898.3155
Chemical Name or Material Dipotassium Glycyrrhizinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.