missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIPK2A Polyclonal Antibody

DIPK2A Polyclonal Antibody
Brand: Invitrogen PA570131
This item is not returnable.
View return policy
Description
DIPK2A Polyclonal Antibody for Western Blot
Specifications
| DIPK2A | |
| Polyclonal | |
| Unconjugated | |
| DIPK2A | |
| 1190002N15Rik; Ab2-095; C3orf58; cc058; chromosome 3 open reading frame 58; deleted in autism 1; deleted in autism protein 1; Deleted in autism protein 1 homolog; DIA1; Dipk2a; Divergent protein kinase domain 2A; Golgi protein GoPro49; Golgi Protein of 49 kDa; GoPro49; HASF; hypoxia and Akt induced stem cell factor; hypoxia and AKT-induced stem cell factor; RGD1309079; RIKEN cDNA 1190002N15 gene; similar to Ab2-095; UPF0672 protein C3orf58; UPF0672 protein C3orf58 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 68861 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3USZ8 | |
| DIPK2A | |
| synthetic peptide directed towards the N-terminal region of mouse 1190002N15Rik. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction