missing translation for 'onlineSavingsMsg'
Learn More

Dipinacolyl methylphosphonate, 97%, Thermo Scientific™

Catalog Number 11415470
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
7040-58-6
C13H29O3P
264.35
MFCD00440711
XSQAITMTGIQAHD-UHFFFAOYNA-N
dipinacolyl methylphosphonate, bis 1,2,2-trimethylpropyl methylphosphonate, bis 3,3-dimethylbutan-2-yl methylphosphonate, phosphonic acid, methyl-, bis 1,2,2-trimethylpropyl ester, methylphosphonic acid bis 1,2,2-trimethylpropyl ester, methylphosphonic acid, bis 1,2,2-trimethylpropyl ester, 3-3,3-dimethylbutan-2-yloxy methyl phosphoryl oxy-2,2-dimet, 3-3,3-dimethylbutan-2-yloxy methyl phosphoryl oxy-2,2-dimethylbutane
23454
3-[3,3-dimethylbutan-2-yloxy(methyl)phosphoryl]oxy-2,2-dimethylbutane
CC(OP(C)(=O)OC(C)C(C)(C)C)C(C)(C)C
This item is not returnable. View return policy
7040-58-6
C13H29O3P
264.35
MFCD00440711
XSQAITMTGIQAHD-UHFFFAOYNA-N
dipinacolyl methylphosphonate, bis 1,2,2-trimethylpropyl methylphosphonate, bis 3,3-dimethylbutan-2-yl methylphosphonate, phosphonic acid, methyl-, bis 1,2,2-trimethylpropyl ester, methylphosphonic acid bis 1,2,2-trimethylpropyl ester, methylphosphonic acid, bis 1,2,2-trimethylpropyl ester, 3-3,3-dimethylbutan-2-yloxy methyl phosphoryl oxy-2,2-dimet, 3-3,3-dimethylbutan-2-yloxy methyl phosphoryl oxy-2,2-dimethylbutane
23454
3-[3,3-dimethylbutan-2-yloxy(methyl)phosphoryl]oxy-2,2-dimethylbutane
CC(OP(C)(=O)OC(C)C(C)(C)C)C(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Boiling Point 75°C (0.8 mmHg)
Quantity 2 g
Sensitivity Air and moisture sensitive
Formula Weight 264.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dipinacolyl methylphosphonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'