missing translation for 'onlineSavingsMsg'
Learn More

Diphosphoryl Chloride 90%, TRC

Product Code. 30378250
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378250 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378250 Supplier Toronto Research Chemicals Supplier No. TRCD49208525G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphosphoryl Chloride
CAS 13498-14-1
Molecular Formula Cl4O3P2
Quantity 25 g
Synonym Pyrophosphoryl Chloride, Phosphorodichloridic Anhydride, Phosphorus Chloride Oxide, Pyrophosphoryl Tetrachloride, Tetrachloropyrophosphoric Acid
InChI Formula InChI=1S/Cl4O3P2/c1-8(2,5)7-9(3,4)6
SMILES ClP(=O)(Cl)OP(=O)(Cl)Cl
Molecular Weight (g/mol) 251.76
Formula Weight 249.8077
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.