missing translation for 'onlineSavingsMsg'
Learn More

Diphosgene, 99%, Thermo Scientific Chemicals

Catalog Number 10032862
Click to view available options
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
503-38-8
C2Cl4O2
197.82
MFCD00015553
HCUYBXPSSCRKRF-UHFFFAOYSA-N
diphosgene, trichloromethyl chloroformate, diphosgen, difosgen czech, perchloromethyl formate, unii-po4q4r80lv, hsdb 371, carbonochloridic acid, trichloromethyl ester, methanol, trichloro-, chloroformate, chloroformic acid trichloromethyl ester
10426
trichloromethyl carbonochloridate
ClC(=O)OC(Cl)(Cl)Cl
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
503-38-8
C2Cl4O2
197.82
MFCD00015553
HCUYBXPSSCRKRF-UHFFFAOYSA-N
diphosgene, trichloromethyl chloroformate, diphosgen, difosgen czech, perchloromethyl formate, unii-po4q4r80lv, hsdb 371, carbonochloridic acid, trichloromethyl ester, methanol, trichloro-, chloroformate, chloroformic acid trichloromethyl ester
10426
trichloromethyl carbonochloridate
ClC(=O)OC(Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
Melting Point -57.0°C
Color Colorless
Density 1.6400g/mL
Boiling Point 128.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCOOCCl3
Refractive Index 1.4560 to 1.4590
Quantity 100 mL
Merck Index 15, 3374
Specific Gravity 1.64
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol, benzene and ether
Formula Weight 197.83
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diphosgene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'