missing translation for 'onlineSavingsMsg'
Learn More

Diphenylzinc, 98+%

Product Code. 11309129
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309129 5 g 5g
11319129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309129 Supplier Thermo Scientific Alfa Aesar Supplier No. 087908.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1078-58-6 | C12H10Zn | 219.59 g/mol

Diphenylzinc is used as the synthetic equivalent of ph- synthon. It is also useful in a Michael 1,4-addition reactions. It is used as a catalyst for propylene oxide polymerization in benzene medium. Further, it is a reactive coupling compound in Negishi cross-coupling reactions. In addition to this, it utilized in the preparation of arylcopper, arylsilver and arylgold compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1078-58-6
Molecular Formula C12H10Zn
Molecular Weight (g/mol) 219.59
MDL Number MFCD00014071
InChI Key BUNROVZNGITPIX-UHFFFAOYSA-N
Synonym diphenylzinc, zinc, diphenyl, mkrvhlwavkjbfn-uhfffaoysa-n, diphenyl zinc, diphenylzink, znph2
PubChem CID 517896
IUPAC Name zinc;benzene
SMILES [Zn++].C1=CC=[C-]C=C1.C1=CC=[C-]C=C1

Specifications

Melting Point 107°C
Boiling Point 280°C to 285°C
Quantity 5 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Soluble in most organic solvents.
Formula Weight 219.58
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material Diphenylzinc

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.