missing translation for 'onlineSavingsMsg'
Learn More

Diphenyltin oxide, Thermo Scientific Chemicals

Product Code. 11324557
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324557 1 g 1g
11304557 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324557 Supplier Thermo Scientific Alfa Aesar Supplier No. 039615.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2273-51-0 | Molecular Formula: C12H10OSn | Molecular Weight: 288.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2273-51-0
Molecular Formula C12H10OSn
Molecular Weight (g/mol) 288.92
MDL Number MFCD00058851
InChI Key VPPWQRIBARKZNY-UHFFFAOYSA-N
Synonym diphenyltin oxide, diphenyltin oxide polymer, stannane, diphenyloxo-, polymer, stannane, oxodiphenyl-, polymer, diphenylstannanone, oxodiphenylstannane, oxo diphenyl stannane, oxo diphenyl tin, stannane, polymer, iv
PubChem CID 75280
IUPAC Name oxo(diphenyl)tin
SMILES O=[Sn](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 320°C (decomposition)
Quantity 1 g
UN Number UN3146
Formula Weight 288.9
Physical Form Powder
Chemical Name or Material Diphenyltin oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.