missing translation for 'onlineSavingsMsg'
Learn More

Diphenyltin Dichloride, TRC

Product Code. 30137812
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137812 10 g 10g
30132909 1 g 1g
17901375 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137812 Supplier Toronto Research Chemicals Supplier No. TRCD49198510G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-99-5
Molecular Formula C12 H10 Cl2 Sn
Molecular Weight (g/mol) 343.82
Synonym Diphenyltin Dichloride, Stannane, dichlorodiphenyl-, Diphenyltin dichloride (6CI), Dichlorodiphenylstannane, Dichlorobis(phenyl)tin, Dichlorodiphenyltin, Diphenyldichlorotin, Diphenylstannyl dichloride, Diphenyltin chloride, NSC 405640
IUPAC Name dichloro(diphenyl)stannane
SMILES Cl[Sn](Cl)(c1ccccc1)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/2C6H5.2ClH.Sn/c2*1-2-4-6-5-3-1;;;/h2*1-5H;2*1H;/q;;;;+2/p-2
Formula Weight 343.9182
Percent Purity >95%
Chemical Name or Material Diphenyl-tin-dichloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.