missing translation for 'onlineSavingsMsg'
Learn More

Diphenylsilanediol, 98%

Product Code. 11404280
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404280 100 g 100g
11494270 25 g 25g
11414280 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404280 Supplier Thermo Scientific Alfa Aesar Supplier No. A10492.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 947-42-2 | C12H12O2Si | 216.311 g/mol

Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

Solubility
Reacts with water.

Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947-42-2
Molecular Formula C12H12O2Si
Molecular Weight (g/mol) 216.311
MDL Number MFCD00002101
InChI Key OLLFKUHHDPMQFR-UHFFFAOYSA-N
Synonym diphenylsilanediol, silanediol, diphenyl, diphenyldihydroxysilane, difenyl-dihydroxysilan, silane, dihydroxydiphenyl, dihydroxy diphenyl silane, diphenylsilandiol, silanediol, 1,1-diphenyl, unii-36x37p8nbb, difenyl-dihydroxysilan czech
PubChem CID 13693
IUPAC Name dihydroxy(diphenyl)silane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(O)O

Specifications

Melting Point ∼150°C (decomposition)
Quantity 100 g
Linear Formula (C6H5Si(OH)2
UN Number UN1325
Beilstein 2523445
Solubility Information Reacts with water.
Formula Weight 216.31
Percent Purity 98%
Odor Odorless
Chemical Name or Material Diphenylsilanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.