missing translation for 'onlineSavingsMsg'
Learn More

Diphenylpyraline Hydrochloride, TRC

Product Code. 30376758
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376758 5 g -
30371306 2.5 g 2.5g
30375377 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376758 Supplier Toronto Research Chemicals Supplier No. TRCD4921135G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-18-3
Molecular Formula C19 H23 N O . Cl H
Molecular Weight (g/mol) 317.85
Synonym Diphenylpyraline Hydrochloride, 1-Methyl-4-piperidyl Benzhydryl Ether Hydrochloride, 4-Diphenylmethoxy-1-methylpiperidine Hydrochloride, Anginosan, Belfene, Dayfen, Diafen, Diafen Hydrochloride, Diaphen, Diphenpyraline Hydrochloride, Diphenylpyraline Teoclate, Hispril, Hispril Hydrochloride, Histryl, Histyn, Kolton Jelly, Lergoban, Lergobine, Sumadil
IUPAC Name 4-benzhydryloxy-1-methylpiperidine;hydrochloride
SMILES Cl.CN1CCC(CC1)OC(c2ccccc2)c3ccccc3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C19H23NO.ClH/c1-20-14-12-18(13-15-20)21-19(16-8-4-2-5-9-16)17-10-6-3-7-11-17;/h2-11,18-19H,12-15H2,1H3;1H
Formula Weight 317.1546
Chemical Name or Material Diphenylpyraline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.