missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylpyraline hydrochloride, MedChemExpress
Description
Diphenylpyraline hydrochloride is a potent histamine H1 receptor antagonist. Diphenylpyraline hydrochloride acts as an orally active antihistamine agent with antimuscarinic and antiallergic effects. Diphenylpyraline hydrochloride can be used for the relief of allergic conditions including rhinitis and hay fever, and in pruritic skin disorders in vivo.
Specifications
Specifications
| Chemical Name or Material | Diphenylpyraline hydrochloride |
| Physical Form | Solid |
| CAS | 132-18-3 |
| Color | White |
| Quantity | 1 g |
| Purity Grade Notes | Research |
| Molecular Formula | C19H24ClNO |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Synonym | 4-Diphenylmethoxy-1-methylpiperidine hydrochloride |
| Solubility Information | H2O : 100 mg/mL (314.61 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (314.61 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?