missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylpyraline Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Diphenylpyraline Hydrochloride |
| CAS | 132-18-3 |
| Analyte or Component Names | Diphenylpyraline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H23 N O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Diphenylpyraline Hydrochloride, 1-Methyl-4-piperidyl Benzhydryl Ether Hydrochloride, 4-Diphenylmethoxy-1-methylpiperidine Hydrochloride, Anginosan, Belfene, Dayfen, Diafen, Diafen Hydrochloride, Diaphen, Diphenpyraline Hydrochloride, Diphenylpyraline Teoclate, Hispril, Hispril Hydrochloride, Histryl, Histyn, Kolton Jelly, Lergoban, Lergobine, Sumadil; |
| InChI Formula | InChI=1S/C19H23NO.ClH/c1-20-14-12-18(13-15-20)21-19(16-8-4-2-5-9-16)17-10-6-3-7-11-17;/h2-11,18-19H,12-15H2,1H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CN1CCC(CC1)OC(c2ccccc2)c3ccccc3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?