missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinyl chloride, 98%

Product Code. 10740721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10740721 50 g Glass bottle 50g
10388630 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10740721 Supplier Thermo Scientific Acros Supplier No. 212960500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1499-21-4 | C12H10ClOP | 236.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-21-4
Molecular Formula C12H10ClOP
Molecular Weight (g/mol) 236.63
MDL Number MFCD00002077
InChI Key QPQGTZMAQRXCJW-UHFFFAOYSA-N
Synonym diphenylphosphinic chloride, diphenylphosphinyl chloride, phosphinic chloride, diphenyl, diphenylphosphinoyl chloride, chlorodiphenylphosphine oxide, dpp-cl, unii-5gp30gy04v, diphenyl phosphinic chloride, diphenylphosphinochloridic acid, diphenylphosphinic acid chloride
PubChem CID 73910
IUPAC Name [chloro(phenyl)phosphoryl]benzene
SMILES ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.2400g/mL
Boiling Point 222.0°C (16.0 mmHg)
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2P(O)Cl
Refractive Index 1.6080 to 1.6120
Quantity 50 g
Beilstein 16, II, 386
Fieser 07,138
Specific Gravity 1.24
Solubility Information Solubility in water: decomposes.
Formula Weight 236.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diphenylphosphinyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diphenylphosphinyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.