missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number 10388630
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1499-21-4
C12H10ClOP
236.63
MFCD00002077
QPQGTZMAQRXCJW-UHFFFAOYSA-N
diphenylphosphinic chloride, diphenylphosphinyl chloride, phosphinic chloride, diphenyl, diphenylphosphinoyl chloride, chlorodiphenylphosphine oxide, dpp-cl, unii-5gp30gy04v, diphenyl phosphinic chloride, diphenylphosphinochloridic acid, diphenylphosphinic acid chloride
73910
[chloro(phenyl)phosphoryl]benzene
ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1499-21-4
C12H10ClOP
236.63
MFCD00002077
QPQGTZMAQRXCJW-UHFFFAOYSA-N
diphenylphosphinic chloride, diphenylphosphinyl chloride, phosphinic chloride, diphenyl, diphenylphosphinoyl chloride, chlorodiphenylphosphine oxide, dpp-cl, unii-5gp30gy04v, diphenyl phosphinic chloride, diphenylphosphinochloridic acid, diphenylphosphinic acid chloride
73910
[chloro(phenyl)phosphoryl]benzene
ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.2400g/mL
Boiling Point 222.0°C (16.0 mmHg)
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2P(O)Cl
Refractive Index 1.6080 to 1.6120
Quantity 10 g
Beilstein 16, II, 386
Fieser 07,138
Specific Gravity 1.24
Solubility Information Solubility in water: decomposes.
Formula Weight 236.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diphenylphosphinyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diphenylphosphinyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'