missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinic Chloride, TRC

Product Code. 30374652
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374652 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374652 Supplier Toronto Research Chemicals Supplier No. TRCD49209510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylphosphinic Chloride
CAS 1499-21-4
Molecular Formula C12H10ClOP
Quantity 10 g
Synonym Chlorodiphenylphosphine Oxide, Diphenylchlorophosphine Oxide, Diphenylphosphinic acid Chloride, Diphenylphosphinic Chloride, Diphenylphosphinoyl Chloride, Diphenylphosphinyl Chloride, Diphenylphosphonic Chloride, Diphenylphosphorus Oxychloride, NSC 175848
InChI Formula InChI=1S/C12H10ClOP/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H
SMILES ClP(=O)(c1ccccc1)c2ccccc2
IUPAC Name [chloro(phenyl)phosphoryl]benzene
Molecular Weight (g/mol) 236.63
Formula Weight 236.0158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.