missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinic Chloride, TRC

Product Code. 30373380
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30373380

Brand: Toronto Research Chemicals TRCD4920955G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylphosphinic Chloride
CAS 1499-21-4
Molecular Formula C12H10ClOP
Quantity 5 g
Synonym Chlorodiphenylphosphine Oxide, Diphenylchlorophosphine Oxide, Diphenylphosphinic acid Chloride, Diphenylphosphinic Chloride, Diphenylphosphinoyl Chloride, Diphenylphosphinyl Chloride, Diphenylphosphonic Chloride, Diphenylphosphorus Oxychloride, NSC 175848
InChI Formula InChI=1S/C12H10ClOP/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H
SMILES ClP(=O)(c1ccccc1)c2ccccc2
IUPAC Name [chloro(phenyl)phosphoryl]benzene
Molecular Weight (g/mol) 236.63
Formula Weight 236.0158
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.