missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinic chloride, 97+%

Product Code. 11499233
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499233 5 g 5g
11409243 25 g 25g
11419243 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499233 Supplier Thermo Scientific Alfa Aesar Supplier No. A11721.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1499-21-4 | C12H10ClOP | 236.63 g/mol

Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.

Solubility
Decomposes with water.

Notes
Moisture Sensitive. Store away from strong bases, moisture. Incompatible with oxidizing agents,, Amines, Alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-21-4
Molecular Formula C12H10ClOP
Molecular Weight (g/mol) 236.63
MDL Number MFCD00002077
InChI Key QPQGTZMAQRXCJW-UHFFFAOYSA-N
Synonym diphenylphosphinic chloride, diphenylphosphinyl chloride, phosphinic chloride, diphenyl, diphenylphosphinoyl chloride, chlorodiphenylphosphine oxide, dpp-cl, unii-5gp30gy04v, diphenyl phosphinic chloride, diphenylphosphinochloridic acid, diphenylphosphinic acid chloride
PubChem CID 73910
IUPAC Name [chloro(phenyl)phosphoryl]benzene
SMILES ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼20°C
Density 1.242
Boiling Point 210°C to 212°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.609
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Decomposes with water.
Formula Weight 236.64
Percent Purity ≥97%
Chemical Name or Material Diphenylphosphinic chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diphenylphosphonic chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Diphenylphosphinic chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.